Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyltrimethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. C19665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19665G Supplier TCI America Supplier No. C19665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 952155-74-7
Molecular Formula C11H20F6N2O4S2
Molecular Weight (g/mol) 422.40
MDL Number MFCD08275363
InChI Key UFYPPIXBMHRHIA-UHFFFAOYSA-N
PubChem CID 44629769
IUPAC Name N,N,N-trimethylcyclohexanaminium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
SMILES C[N+](C)(C)C1CCCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 422.40
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexyltrimethylammonium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.