Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylurea 98.0+%, TCI America™
SDP

Catalog No. C203425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C203425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C203425G Supplier TCI America Supplier No. C203425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 698-90-8
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD00014286
InChI Key WUESWDIHTKHGQA-UHFFFAOYSA-N
Synonym 1-cyclohexylurea, urea, cyclohexyl, n-cyclohexylurea, n-cyclohexlurea, cyclohexyl urea, amino-n-cyclohexylamide, cyclohexyl-urea, monocyclohexylurea, n'-cyclohexylurea, n'-cyclohexyl urea
PubChem CID 69684
IUPAC Name cyclohexylurea
SMILES C1CCC(CC1)NC(=O)N

Specifications

Melting Point 185°C
Color White
Quantity 25 g
Formula Weight 142.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexylurea
RTECSNumber YS7564300
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.