Learn More
Osi Batteries CYCLON 1X6 BATTERY ASSEMBLY
SDP
Catalog No. NC2395965
Encompass_Preferred
Enersys Cyclon 0860-0109 is a 1 x 6 Shrink Wrap battery assembled by OSI Batteries. OSI Batteries is very proud to be an Authorized Distributor and Value Add Center for EnerSys. Please contact us about your OEM Projects.Sealed pure-lead cells were invented by the Gates Corporation in 1973. The purity of the materials used is key to supporting the CYCLON® batterys performance benefits. A longer service life, meaning fewer replacements and the cost associated with it, combined with higher reliability and fewer system failures, result in a lower long-term cost of ownership to the end user or equipment owner.Enersys Cyclon 0860-0109 Monoble Battery General Features:Pure Lead-Tin VRLA AGM DesignSuperior Deep Discharge RecoveryHigh Rate Charge and DischargeTwo Year Shelf Life200+ Full Depth Discharge CyclesOperating Temperature Range -65°C to +80°C
Catalog No. NC2395965 Supplier Osi Batteries Supplier No. 08600109
Enersys Cyclon 0860-0109 is a 1 x 6 Shrink Wrap battery assembled by OSI Batteries. OSI Batteries is very proud to be an Authorized Distributor and Value Add Center for EnerSys. Please contact us about your OEM Projects.Sealed pure-lead cells were invented by the Gates Corporation in 1973. The purity of the materials used is key to supporting the CYCLON® batterys performance benefits. A longer service life, meaning fewer replacements and the cost associated with it, combined with higher reliability and fewer system failures, result in a lower long-term cost of ownership to the end user or equipment owner.Enersys Cyclon 0860-0109 Monoble Battery General Features:Pure Lead-Tin VRLA AGM DesignSuperior Deep Discharge RecoveryHigh Rate Charge and DischargeTwo Year Shelf Life200+ Full Depth Discharge CyclesOperating Temperature Range -65°C to +80°C