Learn More
Perkin Elmer US LLC Cyclonic Injector O-Ring Kit for Avio 200/500
Catalog No. 502112382
Encompass_Preferred
This kit includes: Viton O-Ring 6.07 mm I.D., Qty. 2 (09902207) Viton O-Ring 7.65 mm I.D., Qty. 2 (09926070) PTFE-Coated O-Ring 8.74 mm I.D., Qty. 2 (N0791333)
Catalog No. 50-211-2382 Supplier Perkin Elmer US LLC Supplier No. N0790441
This kit includes: Viton O-Ring 6.07 mm I.D., Qty. 2 (09902207) Viton O-Ring 7.65 mm I.D., Qty. 2 (09926070) PTFE-Coated O-Ring 8.74 mm I.D., Qty. 2 (N0791333)