Learn More
Perkin Elmer US LLC Cyclonic Spray Chamber and Concentric Nebulizer Quick-Change Torch Module for Optima 8x00
Catalog No. 502113149
Encompass_Preferred
This module includes: 1 Slot Quartz Torch (N0780130) MEINHARD Type K1 Nebulizer 1 mL/Min (N0777707) Alumina Injector 2.0 mm I.D. (N0781014) Polyethylene Tubing, 10 ft. (09908265) Adjustable Torch Assembly (N0770600) Cyclonic Spray Chamber with Adjustable Mount (N0770605) Tubing Assembly (N0770610)
Catalog No. 50-211-3149 Supplier Perkin Elmer US LLC Supplier No. N0780607
This module includes: 1 Slot Quartz Torch (N0780130) MEINHARD Type K1 Nebulizer 1 mL/Min (N0777707) Alumina Injector 2.0 mm I.D. (N0781014) Polyethylene Tubing, 10 ft. (09908265) Adjustable Torch Assembly (N0770600) Cyclonic Spray Chamber with Adjustable Mount (N0770605) Tubing Assembly (N0770610)