Learn More
Perkin Elmer US LLC Quartz Baffled Cyclonic Spray Chamber with Gas Port for ELAN 5000/6x00/9000/DRCs PC3 Systems
Catalog No. 509047626
Encompass_Preferred
The quartz baffled cyclonic spray chamber features a 7 mm internal baffle, auxiliary gas port, drain line, and injector connection. It's designed for all-purpose, non-hydrofluoric acid applications. The spray chamber provides excellent sensitivity and stability with low background.
Catalog No. 50-904-7626 Supplier Perkin Elmer US LLC Supplier No. N0777034
The quartz baffled cyclonic spray chamber features a 7 mm internal baffle, auxiliary gas port, drain line, and injector connection. It's designed for all-purpose, non-hydrofluoric acid applications. The spray chamber provides excellent sensitivity and stability with low background.