Learn More
Perkin Elmer US LLC Cyclonic Spray Chamber Injector Support Adapter Kit for Avio 200/500
Catalog No. 502112369
Encompass_Preferred
This kit includes: Cyclonic Concentric Injector Adapter Base Assembly, Qty. 1 (N0790143) Injector Adapter Cap, Qty. 1 (N0791163) Injector Adapter Washer, Qty. 1 (N0776014) Viton O-Ring 6.07 mm I.D., Qty. 1 (09902207) Viton O-Ring 7.65 mm I.D., Qty. 1 (09926070) PTFE Coated O-Ring, 8.74 mm I.D., Qty. 1 (N0791333)
Catalog No. 50-211-2369 Supplier Perkin Elmer US LLC Supplier No. N0790141
This kit includes: Cyclonic Concentric Injector Adapter Base Assembly, Qty. 1 (N0790143) Injector Adapter Cap, Qty. 1 (N0791163) Injector Adapter Washer, Qty. 1 (N0776014) Viton O-Ring 6.07 mm I.D., Qty. 1 (09902207) Viton O-Ring 7.65 mm I.D., Qty. 1 (09926070) PTFE Coated O-Ring, 8.74 mm I.D., Qty. 1 (N0791333)