Learn More
Perkin Elmer US LLC Quartz Baffled Cyclonic PC3 Spray Chamber for Optima 2x00/4x00/5x00/7x00 DV/8x00
Catalog No. 509047628
Encompass_Preferred
The quartz baffled cyclonic spray chamber features a 7 mm internal baffle, drain line, and injector connection. It's designed for all-purpose, non-hydrofluoric acid applications. The spray chamber provides excellent sensitivity, stability and low blanks.
Catalog No. 50-904-7628 Supplier Perkin Elmer US LLC Supplier No. N0777036
The quartz baffled cyclonic spray chamber features a 7 mm internal baffle, drain line, and injector connection. It's designed for all-purpose, non-hydrofluoric acid applications. The spray chamber provides excellent sensitivity, stability and low blanks.