Learn More
Perkin Elmer US LLC Cyclonic/Concentric Sample Introduction Kit for Optima 8x00
Catalog No. 502112256
Encompass_Preferred
This kit includes: 1-Slot Quartz Torch, Qty. 1 (N0780130) 2.0 mm I.D. Alumina Injector, Qty. 1 (N0781014) Glass Cyclonic Spray Chamber, Qty. 1 (N0776052) MEINHARD Type K1 Nebulizer, Qty. 1 (N0777707) Copper Ignitor Tape, 50 Pcs. (N0781097)
Catalog No. 50-211-2256 Supplier Perkin Elmer US LLC Supplier No. N0780617
This kit includes: 1-Slot Quartz Torch, Qty. 1 (N0780130) 2.0 mm I.D. Alumina Injector, Qty. 1 (N0781014) Glass Cyclonic Spray Chamber, Qty. 1 (N0776052) MEINHARD Type K1 Nebulizer, Qty. 1 (N0777707) Copper Ignitor Tape, 50 Pcs. (N0781097)