Learn More
Perkin Elmer US LLC Cyclonic/Concentric Sample Introduction Torch Module for Avio 500
Catalog No. 502112623
Encompass_Preferred
This module kit includes: 0.58 mm I.D. Polyethylene Tubing, 25.4 cm (09908265) Concentric Nebulizer Tubing Assembly (N0770610) Borosilicate Glass MEINHARD Type K1 Concentric Nebulizer (N0777707) 1-Slot Quartz Torch (N0790131) Cyclonic/Concentric Adjustable Injector Assembly (N0790141) Male & Female CTFE Barb Fittings with PFA Capillary Tubing, 0.51 mm I.D. by 25.4 cm Length (N0791018) 2.0 mm I.D. Alumina Injector (N0791183) Baffled Borosilicate Glass Cyclonic Spray Chamber with Drain Fitting (N0791352) Adjustable Torch Assembly Cassette (N0810128)
Catalog No. 50-211-2623 Supplier Perkin Elmer US LLC Supplier No. N0810607
This module kit includes: 0.58 mm I.D. Polyethylene Tubing, 25.4 cm (09908265) Concentric Nebulizer Tubing Assembly (N0770610) Borosilicate Glass MEINHARD Type K1 Concentric Nebulizer (N0777707) 1-Slot Quartz Torch (N0790131) Cyclonic/Concentric Adjustable Injector Assembly (N0790141) Male & Female CTFE Barb Fittings with PFA Capillary Tubing, 0.51 mm I.D. by 25.4 cm Length (N0791018) 2.0 mm I.D. Alumina Injector (N0791183) Baffled Borosilicate Glass Cyclonic Spray Chamber with Drain Fitting (N0791352) Adjustable Torch Assembly Cassette (N0810128)