Learn More
Perkin Elmer US LLC Cyclonic/Concentric Sample Introduction Torch Module for Avio 200
Catalog No. 502113608
Encompass_Preferred
This module kit includes: Baffled Cyclonic Spray Chamber, Qty. 1 (N0791352) 2 mm Alumina Injector, Qty. 1 (N0791183) 1-Slot Quartz Torch, Qty. 1 (N0790131) MEINHARD Type K Nebulizer, Qty. 1 (N0777707) Nebulizer Connector Assembly, Qty. 1 (N0791018) Injector Adapter Kit, Qty. 1 (N0790141) Torch Adapter Kit, Qty. 1 (N0790138) Nebulizer Tubing Assembly, Qty. 1 (N0770610) Polyethylene Tubing 0.58 mm I.D., 25.4 cm Length, Qty. 1 (09908265)
Catalog No. 50-211-3608 Supplier Perkin Elmer US LLC Supplier No. N0790607
This module kit includes: Baffled Cyclonic Spray Chamber, Qty. 1 (N0791352) 2 mm Alumina Injector, Qty. 1 (N0791183) 1-Slot Quartz Torch, Qty. 1 (N0790131) MEINHARD Type K Nebulizer, Qty. 1 (N0777707) Nebulizer Connector Assembly, Qty. 1 (N0791018) Injector Adapter Kit, Qty. 1 (N0790141) Torch Adapter Kit, Qty. 1 (N0790138) Nebulizer Tubing Assembly, Qty. 1 (N0770610) Polyethylene Tubing 0.58 mm I.D., 25.4 cm Length, Qty. 1 (09908265)