Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclooctane

Catalog No. C10940125L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10940125L 2.5 L
C109401100M 100 mL
C109401500M 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C10940125L Supplier Sigma Aldrich Supplier No. C1094012.5L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclooctane

Specifications

CAS 292-64-8
Melting Point 10°C to 13°C (lit.)
Density 0.834 g/mL (at 25°C (literature))
Boiling Point 151°C (740 mmHg)
Molecular Formula C8H16
Refractive Index n20/D 1.458 (literature)
Linear Formula C8H16
MDL Number MFCD00004162
Quantity 2.5 L
Molecular Weight (g/mol) 112.21
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.