Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclooctene Oxide 99%, Thermo Scientific™

Catalog No. AC111451000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC111451000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111451000 Supplier Thermo Scientific Chemicals Supplier No. 111451000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 286-62-4
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C8H14O
Quantity 100g
MDL Number MFCD00005164
Synonym 9-oxabicyclo 6.1.0 nonane, 1,2-epoxycyclooctane, cyclooctene oxide, epoxycyclooctane, cyclooctene, oxide, cyclooctane, 1,2-epoxy, ccris 3753, 9-oxabicyclo 6.1.0 nonane, cis, cyclooctane epoxide, cyclooctane,2-epoxy
InChI Key MELPJGOMEMRMPL-UHFFFAOYSA-N
SMILES C1CCCC2C(O2)CC1
IUPAC Name 9-oxabicyclo[6.1.0]nonane
Molecular Weight (g/mol) 126.199
PubChem CID 67513
Formula Weight 126.2
Percent Purity ≥98.5%
Chemical Name or Material Cyclooctene oxide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Flammable solid.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-010-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.