Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclooctylamine, 97+%, Thermo Scientific™

Catalog No. AAL0098106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0098106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0098106 Supplier Thermo Scientific Chemicals Supplier No. L0098106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cyclooctylamine is used as pharmaceutical intermediates.

Solubility
Soluble in water. 10.2 g/L at 20°C.

Notes
Protect from moisture, heat. Avoid excessive and light exposure. Air sensitive. Store at +15°C to +25°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5452-37-9
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00001748
InChI Key HSOHBWMXECKEKV-UHFFFAOYSA-N
Synonym cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc
PubChem CID 2903
IUPAC Name cyclooctanamine
SMILES C1CCCC(CCC1)N

Specifications

Melting Point -48°C
Density 0.912
Boiling Point 189°C to 190°C
Flash Point 62°C (143°F)
Odor Amine-like
Refractive Index 1.481
Quantity 5 g
UN Number UN2735
Beilstein 2689120
Sensitivity Air Sensitive
Solubility Information Soluble in water. 10.2g/L at 20°C.
Formula Weight 127.23
Percent Purity ≥97%
Chemical Name or Material Cyclooctylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 226-694-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.