Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclooctyne-o-amido-peg4-vc-pab-gly-gly-nh-o-co-exatecan | 2699066-62-9 | 97.3% | 1396.51 | C69H90FN11O19 | 1 MG
SDP

Catalog No. 5000204440
Encompass_Preferred

Cyclooctyne-O-amido-PEG4-VC-PAB-Gly-Gly-NH-O-CO-Exatecan is a cleavable drug-linker conjugate intended for antibody-drug conjugate (ADC) research, where Exatecan serves as the cytotoxic payload. Supplied as a solid for research use only, it is used in conjugation chemistry and ADC synthesis.

  • Molecular formula: C69H90FN11O19.
  • Molecular weight: 1396.51.
  • Purity: 97.3%.
  • Physical state: solid.
  • Cas number: 2699066-62-9.
  • Available sizes: 1 mg and 5 mg vials; larger quantities available by quotation.
  • Storage: 4°C under nitrogen; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month), stored under nitrogen.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-04440 Supplier Medchemexpress LLC Supplier No. HY1574071MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclooctyne-O-amido-PEG4-VC-PAB-Gly-Gly-NH-O-CO-Exatecan is a cleavable drug-linker conjugate intended for antibody-drug conjugate (ADC) research, where Exatecan serves as the cytotoxic payload. Supplied as a solid for research use only, it is used in conjugation chemistry and ADC synthesis.

  • Molecular formula: C69H90FN11O19.
  • Molecular weight: 1396.51.
  • Purity: 97.3%.
  • Physical state: solid.
  • Cas number: 2699066-62-9.
  • Available sizes: 1 mg and 5 mg vials; larger quantities available by quotation.
  • Storage: 4°C under nitrogen; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month), stored under nitrogen.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.