Learn More
Medchemexpress LLC Cyclooctyne-O-amido-PEG4-VC-PAB-Gly-Gly-NH-O-CO-Exatecan | 97.3% | 1396.51 Da | 5 MG
SDP
Catalog No. 5000204505
Encompass_Preferred
Cyclooctyne-O-amido-PEG4-VC-PAB-Gly-Gly-NH-O-CO-Exatecan is a drug-linker conjugate designed for antibody-drug conjugate (ADC) research; the construct contains an exatecan payload, a PEG4-VC-PAB cleavable linker, and a cyclooctyne reactive group for conjugation.
  • Drug-linker conjugate for ADC research.
  • Contains exatecan as the cytotoxic payload.
  • Cyclooctyne reactive group suitable for bioorthogonal conjugation.
  • PEG4-VC-PAB linker provides a cleavable spacer for payload release.
  • Supplied at high purity suitable for research applications.
  • Available in small pack sizes for early-stage research and conjugation development.
Catalog No. 50-002-04505 Supplier Medchemexpress LLC Supplier No. HY1574075MG
Cyclooctyne-O-amido-PEG4-VC-PAB-Gly-Gly-NH-O-CO-Exatecan is a drug-linker conjugate designed for antibody-drug conjugate (ADC) research; the construct contains an exatecan payload, a PEG4-VC-PAB cleavable linker, and a cyclooctyne reactive group for conjugation.
  • Drug-linker conjugate for ADC research.
  • Contains exatecan as the cytotoxic payload.
  • Cyclooctyne reactive group suitable for bioorthogonal conjugation.
  • PEG4-VC-PAB linker provides a cleavable spacer for payload release.
  • Supplied at high purity suitable for research applications.
  • Available in small pack sizes for early-stage research and conjugation development.