Learn More
Medchemexpress LLC Cyclooctyne-O-PFP ester | 886209-60-5 | 95.1% | 348.26 | C16H13F5O3 | 25 MG
SDP
Catalog No. 5000202948
Encompass_Preferred
Cyclooctyne-O-PFP ester is a cleavable ADC linker and click-chemistry reagent containing an alkyne functional group used to conjugate azide-containing molecules via azide-alkyne cycloaddition. It is supplied as a solid for research use and is commonly applied in bioconjugation and antibody-drug conjugate synthesis.
  • Cleavable ADC linker for bioconjugation and antibody-drug conjugate synthesis.
  • Reacts in copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Supplied in small quantities suitable for research use and method development.
  • High purity suitable for conjugation chemistry, with a molecular weight of 348.26 g·mol⁻¹.
Catalog No. 50-002-02948 Supplier Medchemexpress LLC Supplier No. HY12651825MG
Cyclooctyne-O-PFP ester is a cleavable ADC linker and click-chemistry reagent containing an alkyne functional group used to conjugate azide-containing molecules via azide-alkyne cycloaddition. It is supplied as a solid for research use and is commonly applied in bioconjugation and antibody-drug conjugate synthesis.
  • Cleavable ADC linker for bioconjugation and antibody-drug conjugate synthesis.
  • Reacts in copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Supplied in small quantities suitable for research use and method development.
  • High purity suitable for conjugation chemistry, with a molecular weight of 348.26 g·mol⁻¹.