Learn More
Enzo Life Sciences Cyclooxygenase (sheep), (purified) (1mg)
SDP
Catalog No. 502010542
Encompass_Preferred
Alternative name: COX. Purity: ≥95%. Formulation: Liquid. In 0.1M Tris-HCl, pH 7.4 containing 1mM Tryptophan and 1% Tween-80. Source: Isolated from ram seminal vesicles. Long Term Storage: -80°C.
Catalog No. 50-201-0542 Supplier Enzo Life Sciences Supplier No. BMLEH0010001
Alternative name: COX. Purity: ≥95%. Formulation: Liquid. In 0.1M Tris-HCl, pH 7.4 containing 1mM Tryptophan and 1% Tween-80. Source: Isolated from ram seminal vesicles. Long Term Storage: -80°C.