Learn More
Medchemexpress LLC HY-17024 100mg , Cyclopamine CAS:4449-51-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17024100MG
Encompass_Preferred
HY-17024 100mg Cyclopamine CAS: 4449-51-8 Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0995