Learn More
BIOGEMS INTERNATIONAL INC Cyclopamine 10mg
SDP
Catalog No. 501530147
Encompass
Cyclopamine is a steroidal alkaloid inhibitor of the hedgehog signaling pathway (Hh), an essential pathway for embryonic development. It is reported to be capable of inducing differentiation of stem cells into endocrine cells and various anti-cancer applications by suppressing excess Hh activity.
Catalog No. 50-153-0147 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 444518510MG
Cyclopamine is a steroidal alkaloid inhibitor of the hedgehog signaling pathway (Hh), an essential pathway for embryonic development. It is reported to be capable of inducing differentiation of stem cells into endocrine cells and various anti-cancer applications by suppressing excess Hh activity.