Learn More
Cayman Chemical Cyclopamine, 4449-51-8, 10 mg
SDP
Supplier:  Cayman Chemical 1132110
Encompass_Preferred
Molecular Formula C27H41NO2, Purity ≥95%, Formula Weight 411.6, Synonyms: (2'R,3S,3'R,3'aS,6'S,6aS,6bS,7'aR,11aS,11bR)-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3',6',10,11b-tetramethyl-spiro[9H-benzo[a]fluorene-9,2'(3'H)-furo[3,2-b]pyridin]-3-ol, 11-Deoxojervine, Jervine; A natural steroidal alkaloid that inhibits signaling through the Hh pathway at the level of the pathway activator Smo; inhibits Hh-dependent expression of Pax7 with an IC50 value of 24 nM; has potential applications in the treatment of cancer.
Catalog No. NC0730748