Learn More
Selleck Chemical LLC Cyclopamine 5mg 4449-51-8 11-deoxojervine
SDP
Supplier:  Selleck Chemical LLC S11465MG
Encompass_Preferred
Cyclopamine (11-deoxojervine) is a specific Hedgehog (Hh) signaling pathway antagonist of Smoothened (Smo) with IC50 of 46 nM in TM3Hh12 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-760-4