Learn More
Selleck Chemical LLC Cyclopamine 50mg 4449-51-8 11-deoxojervine
SDP
Catalog No. 507607
Encompass_Preferred
Cyclopamine (11-deoxojervine) is a specific Hedgehog (Hh) signaling pathway antagonist of Smoothened (Smo) with IC50 of 46 nM in TM3Hh12 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-760-7 Supplier Selleck Chemical LLC Supplier No. S114650MG
Cyclopamine (11-deoxojervine) is a specific Hedgehog (Hh) signaling pathway antagonist of Smoothened (Smo) with IC50 of 46 nM in TM3Hh12 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.