Learn More
Cayman Chemical Cyclopamine, 4449-51-8, 1 mg
SDP
Supplier:  Cayman Chemical 113211
Encompass_Preferred
A natural steroidal alkaloid that inhibits signaling through the Hh pathway at the level of the pathway activator Smo; inhibits Hh-dependent expression of Pax7 with an IC50 value of 24 nM; has potential applications in the treatment of cancer
Catalog No. NC1117387