Learn More
Apexbio Technology LLC Cyclopamine 4449-51-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062214
Encompass_Preferred
Cyclopamine (CAS 4449-51-8) is a naturally occurring steroidal alkaloid that selectively inhibits the Hedgehog (Hh) signaling pathway a conserved cascade involved in cellular proliferation and differentiation In human breast cancer cell lines (MCF-7 and MDA-MB-231) cyclopamine treatment (10 20 M) significantly suppresses proliferation induces cell cycle arrest at G1 phase and reduces invasive capabilities In FXR-bla assays cyclopamine exhibits an EC50 of 10 57 M As an Hh-antagonist it is widely utilized in research examining embryogenesis cancer biology tumor invasiveness and developmental defects
Catalog No. 50-000-62214 Supplier Apexbio Technology LLC Supplier No. A83405.1
Cyclopamine (CAS 4449-51-8) is a naturally occurring steroidal alkaloid that selectively inhibits the Hedgehog (Hh) signaling pathway a conserved cascade involved in cellular proliferation and differentiation In human breast cancer cell lines (MCF-7 and MDA-MB-231) cyclopamine treatment (10 20 M) significantly suppresses proliferation induces cell cycle arrest at G1 phase and reduces invasive capabilities In FXR-bla assays cyclopamine exhibits an EC50 of 10 57 M As an Hh-antagonist it is widely utilized in research examining embryogenesis cancer biology tumor invasiveness and developmental defects