Learn More
Medchemexpress LLC Cyclopamine | 4449-51-8 | 411.62 | 200 MG
SDP
Catalog No. 5000385709
Encompass_Preferred
Cyclopamine is a steroidal alkaloid and a selective Smo inhibitor that acts as a Hedgehog (Hh) pathway antagonist, with an IC50 of 46 nM in Hh cell assays. It is used in research for its ability to suppress cell growth and inhibit Hh signaling.
  • Hedgehog (Hh) pathway antagonist
  • Selective Smo inhibitor
  • Suppresses cell growth
  • Inhibits Hh signaling through direct interaction with Smo
  • Suitable for research and analytical applications
  • Induces tumor remission in medulloblastoma models
  • Blocks tumor formation in pancreatic adenocarcinoma cells
Catalog No. 50-003-85709 Supplier Medchemexpress LLC Supplier No. HY17024200MG
Cyclopamine is a steroidal alkaloid and a selective Smo inhibitor that acts as a Hedgehog (Hh) pathway antagonist, with an IC50 of 46 nM in Hh cell assays. It is used in research for its ability to suppress cell growth and inhibit Hh signaling.
  • Hedgehog (Hh) pathway antagonist
  • Selective Smo inhibitor
  • Suppresses cell growth
  • Inhibits Hh signaling through direct interaction with Smo
  • Suitable for research and analytical applications
  • Induces tumor remission in medulloblastoma models
  • Blocks tumor formation in pancreatic adenocarcinoma cells