Learn More
Medchemexpress LLC HY-17024 5mg Medchemexpress, Cyclopamine CAS:4449-51-8 Purity:>98%
SDP
Catalog No. 502029228
Encompass_Preferred
Medchemexpress, HY-17024 5mg Cyclopamine CAS:4449-51-8 Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9228 Supplier Medchemexpress LLC Supplier No. HY170245MG
Medchemexpress, HY-17024 5mg Cyclopamine CAS:4449-51-8 Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.