Learn More
Cayman Chemical Cyclopamn 10mg
SDP
Catalog No. 502951730
Encompass_Preferred
A natural steroidal alkaloid that inhibits signaling through the Hh pathway at the level of the pathway activator Smo; inhibits Hh-dependent expression of Pax7 with an IC50 value of 24 nM; has potential applications in the treatment of cancer
Catalog No. 50-295-1730 Supplier Cayman Chemical Supplier No. 1132110 MG
A natural steroidal alkaloid that inhibits signaling through the Hh pathway at the level of the pathway activator Smo; inhibits Hh-dependent expression of Pax7 with an IC50 value of 24 nM; has potential applications in the treatment of cancer