Learn More
AdipoGen Cyclopenin
SDP
Catalog No. 502562654
Encompass_Preferred
Chemical. CAS 19553-26-5 and 20007-87-8. Formula C17H14N2O3. MW 294.3. Isolated from Penicillium sp. Antimicrobial. Phytotoxic. Specific acetylcholinesterase AChE inhibitor.
Catalog No. 50-256-2654 Supplier AdipoGen Supplier No. AGCN20134M001
Chemical. CAS 19553-26-5 and 20007-87-8. Formula C17H14N2O3. MW 294.3. Isolated from Penicillium sp. Antimicrobial. Phytotoxic. Specific acetylcholinesterase AChE inhibitor.