Learn More
AdipoGen Cyclopenin (5 mg)
SDP
Supplier:  AdipoGen AGCN20134M005
Encompass_Preferred
Chemical. CAS: 19553-26-5 and 20007-87-8. Formula: C17H14N2O3. MW: 294.3. Isolated from Penicillium sp. Antimicrobial. Phytotoxic. Specific acetylcholinesterase (AChE) inhibitor.
Catalog No. 50-205-3509