Learn More
Medchemexpress LLC Cyclopent[b]indol-1(2H)-one, 3,4-dihydro-4-methyl- | 50776-26-6 | 99.9% | C12H11NO | 500 MG

Supplier: Medchemexpress LLC HY75357500MG
4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one, also identified as Cyclopent[b]indol-1(2H)-one, 3,4-dihydro-4-methyl-, is a chemical compound primarily used as a drug intermediate. It is critical for the synthesis of various active compounds in research settings.
- Used as a drug intermediate for synthesis
- Intended for research use only
- Not fully validated for medical applications
- Exhibits a purity of 99.94%
- Molecular formula is C12H11NO
- Molecular weight is 185.23
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.