Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadecanone, 99%, Thermo Scientific™

Catalog No. AAA1765203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1765203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1765203 Supplier Thermo Scientific Chemicals Supplier No. A1765203

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 502-72-7
Melting Point 62°C to 66°C
Density 0.897
Boiling Point 120°C (0.3 mmHg)
Molecular Formula C15H28O
Quantity 1 g
MDL Number MFCD00001266
Beilstein 1618444
Synonym exaltone, normuscone, normuscon, unii-t09g246let, exalton, cyclopentadecanon, cylcopentadecanone, cyclopentandecanone, cyclopentadecan-1-one, cyclopentadecanone
InChI Key OSOIQJGOYGSIMF-UHFFFAOYSA-N
SMILES C1CCCCCCCC(=O)CCCCCC1
IUPAC Name cyclopentadecanone
Molecular Weight (g/mol) 224.388
PubChem CID 10409
Formula Weight 224.39
Percent Purity 99%
Chemical Name or Material Cyclopentadecanone
Show More Show Less
EINECSNumber 207-951-2
RTECSNumber GY0900000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.