Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopentadienyl Iron(II) Dicarbonyl Dimer

Catalog No. 22548725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
125 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-548-725G 25 g
22-548-7125G 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-548-725G Supplier Sigma Aldrich Supplier No. 22548725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclopentadienyl Iron(II) Dicarbonyl Dimer

Specifications

CAS 12154-95-9
Melting Point 194°C
Molecular Formula C14H10Fe2O4
Linear Formula C14H10Fe2O4
Quantity 25 g
MDL Number MFCD00001436
Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyliron dicarbonyl dimer; Tetracarbonylbis(cyclopentadienyl)diiron; Bis(cyclopentadienyl)tetracarbonyl-diiron; Di(cyclopentadienyl)tetracarbonyl-diiron
Molecular Weight (g/mol) 353.92 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.