Learn More
Strem, An Ascensus Company CAS# 12079-65-1. 1g. Cyclopentadienylmanganese tricarbonyl, 98% Cymantrene. MFCD00001440
SDP
Catalog No. 501385447
Encompass
CAS# 12079-65-1. 1g. Cyclopentadienylmanganese tricarbonyl, 98% Cymantrene. MFCD00001440. Molecular Weight: 204.06. Molecular Formula: C5H5Mn(CO)3. Color/form: yellow xtl. Strem# 25-0390. http://www.strem.com/catalog/v/25-0390/
Catalog No. 50-138-5447 Supplier Strem, An Ascensus Company Supplier No. 2503901G
CAS# 12079-65-1. 1g. Cyclopentadienylmanganese tricarbonyl, 98% Cymantrene. MFCD00001440. Molecular Weight: 204.06. Molecular Formula: C5H5Mn(CO)3. Color/form: yellow xtl. Strem# 25-0390. http://www.strem.com/catalog/v/25-0390/