Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienyliron Dicarbonyl Dimer 96.0+%, TCI America™
SDP

Catalog No. C15925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15925G Supplier TCI America Supplier No. C15925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12154-95-9
Molecular Formula C14H10Fe2O4
MDL Number MFCD00001436
Synonym Bis(cyclopentadienyl)tetracarbonyl Diiron

Specifications

Quantity 5 g
Formula Weight 353.92
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentadienyliron Dicarbonyl Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.