Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienylmanganese tricarbonyl

Catalog No. AA3860614
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3860614 25 g
AA3860603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3860614 Supplier Thermo Scientific Chemicals Supplier No. 3860614

CAS: 12079-65-1

Cyclopentadienylmanganese tricarbonyl is an important raw material and intermediate used in organic synthesis, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12079-65-1
MDL Number MFCD00001440
Synonym Tricarbonylcyclopentadienylmanganese

Specifications

Melting Point 77°C
Physical Form Crystalline
Quantity 25 g
UN Number UN3466
Formula Weight 204.06
Sensitivity Air Sensitive
Chemical Name or Material Cyclopentadienylmanganese tricarbonyl
Hazard Category Danger
Hazard Statement GHS H Statement H300-H330 Fatal if swallowed. Fatal if inhaled.
Precautionary Statement GHS P Statement P260-P301+P310-P304+P340-P320-P330-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Specific treatment is urgent (see label). Rinse mouth. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.