Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienylniobium(V) tetrachloride, Thermo Scientific Chemicals

Catalog No. AA39246MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39246MD 250 mg
AA3924606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39246MD Supplier Thermo Scientific Chemicals Supplier No. 039246MD
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33114-15-7 | Molecular Formula: C5H5Cl4Nb | Molecular Weight: 299.801 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33114-15-7
Molecular Formula C5H5Cl4Nb
Molecular Weight (g/mol) 299.801
MDL Number MFCD00070445
InChI Key KAWQZSRKEBAOSY-UHFFFAOYSA-J
Synonym niobium 5+ chloride cyclopenta-1,3-dien-1-ide 1/4/1, niobium 5+ ion cyclopenta-1,3-dien-1-ide tetrachloride
PubChem CID 74765428
IUPAC Name cyclopenta-1,3-diene;niobium(5+);tetrachloride
SMILES C1C=CC=[C-]1.[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]

Specifications

Melting Point 180°C (decomposition)
Color Yellow to Brown
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 299.82
Physical Form Crystalline
Chemical Name or Material Cyclopentadienylniobium(V) tetrachloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.