Learn More
Strem, An Ascensus Company CAS# 1270-98-0. 25g. Cyclopentadienyltitanium trichloride, 99%. MFCD00003724
SDP
Catalog No. 5090110743
Encompass
CAS# 1270-98-0. 25g. Cyclopentadienyltitanium trichloride, 99%. MFCD00003724. Molecular Weight: 219.35. Molecular Formula: C5H5TiCl3. Color/form: yellow xtl. Strem# 22-0400. http://www.strem.com/catalog/v/22-0400/
Catalog No. 50-901-10743 Supplier Strem, An Ascensus Company Supplier No. 22040025G
CAS# 1270-98-0. 25g. Cyclopentadienyltitanium trichloride, 99%. MFCD00003724. Molecular Weight: 219.35. Molecular Formula: C5H5TiCl3. Color/form: yellow xtl. Strem# 22-0400. http://www.strem.com/catalog/v/22-0400/