Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020679 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0679 5 mL
11-102-0620 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0679 Supplier MilliporeSigma Supelco Supplier No. 296805ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopentane is a colourless flammable acyclic hydrocarbon liquid. It is a widely used component in preparing products like analgesics, insecticides, sedatives, antitumor agents and also finds application in pharmaceutical industry.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopentane
CAS 287-92-3
Ignition Point 682°F
Vapor Density 2 (vs. air)
Density 0.751 g/mL (at 25°C)
Flash Point -20°C - Closed Cup
Physical Form Neat
Linear Formula C5H10
Technique Gas Chromatography
MDL Number MFCD00001356
Melting Point -94°C
Boiling Point 50°C
Type Analytical Standard
Industry Type Environmental Testing
Molecular Weight (g/mol) 70.13
Quantity 5 mL
Formula Weight 70.13
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.