Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclopentaneacetic acid, 3-oxo-2-pentyl- | 3572-64-3 | 99.3% | 250 MG
SDP

Catalog No. 5000341776
Encompass_Preferred

Dihydrojasmonic acid acts as a plant growth regulator. This product is intended for research use only and has not been fully validated for medical applications.

  • Appears as a colorless to light yellow liquid.
  • Purity of 99.3%.
  • Molecular weight of 212.29.
  • Derived from plants.

Catalog No. 50-003-41776 Supplier Medchemexpress LLC Supplier No. 250MGHY131116
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrojasmonic acid acts as a plant growth regulator. This product is intended for research use only and has not been fully validated for medical applications.

  • Appears as a colorless to light yellow liquid.
  • Purity of 99.3%.
  • Molecular weight of 212.29.
  • Derived from plants.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.