Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopentanecarboxaldehyde

Catalog No. 5260375G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-603-75G 5 g
52-603-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-603-75G Supplier Sigma Aldrich Supplier No. 5260375G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclopentanecarboxaldehyde can be prepared from cyclohexene, via copper-catalyzed oxidation with persulfate. it can also be synthesized from mercuric sulfate and cyclohexene.cyclopentanecarboxaldehyde may be used in the synthesis of:
  • D,L-trans-2-cyclopentylmethylene-1-nitroso-1-(2-hydroxycyclohexyl)hydrazine
  • Ethyl 5-cyclopentyl-5-hydroxy-2-pentenoate
  • Ethyl 3-cyclo entyl-3-hydroxy-2-vinylpropanoate

Specifications

CAS 872-53-7
Density 0.919 g/mL (at 25°C (literature))
Boiling Point 140°C to 141°C (lit.)
Molecular Formula C6H10O
Refractive Index n20/D 1.4430 (literature)
Linear Formula C5H9CHO
MDL Number MFCD00060798
Quantity 5 g
Molecular Weight (g/mol) 98.14
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.