Learn More
Medchemexpress LLC Cyclopentanecarboxylic acid, 1-(4-nitrophenyl)-, 2-(diethylamino)ethyl ester, hydrochloride | 98636-73-8 | MFCD00673872 | 99.6% | 370.87 g/mol | C18H27ClN2O4 | 25 MG
SDP
Catalog No. 5000212595
Encompass_Preferred
Nitrocaramiphen hydrochloride is a selective M1 muscarinic receptor antagonist supplied as the hydrochloride salt for research use. It inhibits muscarine-induced hyperpolarization in muscle fibers and is provided as a white to off-white solid suitable for biochemical and pharmacological studies.
  • Selective M1 receptor antagonist (Ki: 5.5 nM).
  • High purity: 99.6% (manufacturer COA).
  • White to off-white solid, molecular formula C18H27ClN2O4.
  • Molecular weight 370.87 g/mol.
  • Available in small milligram quantities for research use.
  • Store sealed away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-12595 Supplier Medchemexpress LLC Supplier No. HY10094525MG
Nitrocaramiphen hydrochloride is a selective M1 muscarinic receptor antagonist supplied as the hydrochloride salt for research use. It inhibits muscarine-induced hyperpolarization in muscle fibers and is provided as a white to off-white solid suitable for biochemical and pharmacological studies.
  • Selective M1 receptor antagonist (Ki: 5.5 nM).
  • High purity: 99.6% (manufacturer COA).
  • White to off-white solid, molecular formula C18H27ClN2O4.
  • Molecular weight 370.87 g/mol.
  • Available in small milligram quantities for research use.
  • Store sealed away from moisture; in solvent: -80°C (6 months), -20°C (1 month).