Learn More
Medchemexpress LLC Cyclopentanecarboxylic acid, 3-[[(1,1-dimethylethoxy)carbonyl)amino]-, (1S,3R)- | 261165-05-3 | 229.27 | 5 G
SDP
Catalog No. 5000447357
Encompass_Preferred
BOC-(1S,3R)-3-aminocyclopentanecarboxylic acid is a synthetic intermediate primarily used in pharmaceutical synthesis. This product is intended for research purposes only and not for patient use.
  • Useful for pharmaceutical synthesis
  • Appears as a white to off-white solid
  • Purity of 98.0%
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years and -20°C for 1 year
Catalog No. 50-004-47357 Supplier Medchemexpress LLC Supplier No. HYW0062025G
BOC-(1S,3R)-3-aminocyclopentanecarboxylic acid is a synthetic intermediate primarily used in pharmaceutical synthesis. This product is intended for research purposes only and not for patient use.
  • Useful for pharmaceutical synthesis
  • Appears as a white to off-white solid
  • Purity of 98.0%
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years and -20°C for 1 year