Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020337 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0337 5 mL
11-101-8514 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0337 Supplier MilliporeSigma Supelco Supplier No. 082995MLF
Add to Cart
Add to Cart

Cyclopentanone is one of the industrially important compounds employed in the synthesis of fungicides, pharmaceuticals, rubber chemicals and perfume products.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 120-92-3
Density 0.951 g/mL (at 25°C)
Flash Point 30°C - Closed Cup
Linear Formula C5H8(=O)
MDL Number MFCD00001409
Melting Point -51°C
Boiling Point 130°C to 131°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 84.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.