Learn More
Medchemexpress LLC Cyclopentenyl uracil | 90597-20-9 | 99.1% | 240.22 g·mol⁻1 | C10H12N2O5 | 5 MG
SDP
Catalog No. 5000221089
Encompass_Preferred
Cyclopentenyl uracil is a non-cytotoxic uridine kinase inhibitor used in biochemical and in vivo research to block the salvage of circulating uridine by host and tumor tissues. It is supplied as a solid with reported high purity and is offered in small research pack sizes and DMSO solution formats for flexible dosing in cellular and animal studies.
  • Inhibits uridine kinase activity in biochemical assays.
  • Blocks uridine salvage in vivo to support metabolism studies.
  • High reported purity suitable for research applications.
  • Available as a solid and as a DMSO solution for dosing flexibility.
  • Well-characterized small-molecule properties for experimental reproducibility.
Catalog No. 50-002-21089 Supplier Medchemexpress LLC Supplier No. HY1456685MG
Cyclopentenyl uracil is a non-cytotoxic uridine kinase inhibitor used in biochemical and in vivo research to block the salvage of circulating uridine by host and tumor tissues. It is supplied as a solid with reported high purity and is offered in small research pack sizes and DMSO solution formats for flexible dosing in cellular and animal studies.
  • Inhibits uridine kinase activity in biochemical assays.
  • Blocks uridine salvage in vivo to support metabolism studies.
  • High reported purity suitable for research applications.
  • Available as a solid and as a DMSO solution for dosing flexibility.
  • Well-characterized small-molecule properties for experimental reproducibility.