Learn More
Medchemexpress LLC HY-B1621A 10mg Medchemexpress, Cyclopentolate (hydrochloride) CAS:5870-29-1 Purity:>98%
SDP
Catalog No. 502031211
Encompass_Preferred
Medchemexpress, HY-B1621A 10mg Cyclopentolate (hydrochloride) CAS:5870-29-1 Cyclopentolate (DL-Cyclopentolate) hydrochloride is an Atropine-like muscarinic receptors antagonist with a pKB value of 7.8 (on the circular ciliary muscle). Cyclopentolate hydrochloride is an anti-muscarinic agent commonly used in the ophthalmologic practice[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1211 Supplier Medchemexpress LLC Supplier No. HYB1621A10MG
Medchemexpress, HY-B1621A 10mg Cyclopentolate (hydrochloride) CAS:5870-29-1 Cyclopentolate (DL-Cyclopentolate) hydrochloride is an Atropine-like muscarinic receptors antagonist with a pKB value of 7.8 (on the circular ciliary muscle). Cyclopentolate hydrochloride is an anti-muscarinic agent commonly used in the ophthalmologic practice[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.