Learn More
Selleck Chemical LLC Cyclopentolate Hydrochloride
SDP
Catalog No. 5000003908
Encompass_Preferred
Cyclopentolate is an antagonist of muscarinic acetylcholine receptors with Ki values of 1 62 27 5 and 2 63 nM for M1 M2 and M3 receptors respectively
Catalog No. 50-000-03908 Supplier Selleck Chemical LLC Supplier No. S592125MG
Cyclopentolate is an antagonist of muscarinic acetylcholine receptors with Ki values of 1 62 27 5 and 2 63 nM for M1 M2 and M3 receptors respectively