Learn More
Medchemexpress LLC Cyclopentolate (hydrochloride) | 5870-29-1 | 1 ML
SDP
Catalog No. 5000413291
Encompass_Preferred
Cyclopentolate hydrochloride is an atropine-like muscarinic receptor antagonist with a pKB value of 7.8 (on the circular ciliary muscle). It is an anti-muscarinic agent commonly used in ophthalmologic practice.
  • Antagonist of atropine-like muscarinic receptors
  • pKB value of 7.8 on the circular ciliary muscle
  • Used as an anti-muscarinic agent
  • Commonly used in ophthalmologic practice
Catalog No. 50-004-13291 Supplier Medchemexpress LLC Supplier No. HYB1621A10MM/1ML
Cyclopentolate hydrochloride is an atropine-like muscarinic receptor antagonist with a pKB value of 7.8 (on the circular ciliary muscle). It is an anti-muscarinic agent commonly used in ophthalmologic practice.
  • Antagonist of atropine-like muscarinic receptors
  • pKB value of 7.8 on the circular ciliary muscle
  • Used as an anti-muscarinic agent
  • Commonly used in ophthalmologic practice