Learn More
Cayman Chemical CyclopentolatehydroCLOrID 10mg
SDP
Catalog No. 502994418
Encompass_Preferred
A muscarinic receptor antagonist (Kis = 1.62, 27.5, and 2.63 nM for M1, M2, and M3 receptors, respectively); inhibits carbachol-induced contraction of isolated human iris sphincter, circular ciliary muscle, and longitudinal ciliary muscle (Kbs = 7.9, 15.8, and 12.5 nM, respectively)
Catalog No. 50-299-4418 Supplier Cayman Chemical Supplier No. 2394410 MG
A muscarinic receptor antagonist (Kis = 1.62, 27.5, and 2.63 nM for M1, M2, and M3 receptors, respectively); inhibits carbachol-induced contraction of isolated human iris sphincter, circular ciliary muscle, and longitudinal ciliary muscle (Kbs = 7.9, 15.8, and 12.5 nM, respectively)