Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclopentyl-boronic acid | 63076-51-7 | 113.95 | 10 G
SDP

Catalog No. 5000446332
Encompass_Preferred

Cyclopentylboronic acid is a biochemical reagent used in life science research. It functions as both a biological material and an organic compound.

  • Suitable as a biological material or organic compound
  • White to off-white solid appearance
  • Stable for 3 years as a powder when stored at -20°C
  • Stable for 6 months in solvent when stored at -80°C
  • Stable for 1 month in solvent when stored at -20°C

Catalog No. 50-004-46332 Supplier Medchemexpress LLC Supplier No. HYW00095510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopentylboronic acid is a biochemical reagent used in life science research. It functions as both a biological material and an organic compound.

  • Suitable as a biological material or organic compound
  • White to off-white solid appearance
  • Stable for 3 years as a powder when stored at -20°C
  • Stable for 6 months in solvent when stored at -80°C
  • Stable for 1 month in solvent when stored at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.